Learn More
Enzo Life Sciences MK-571 . sodium salt (5 mg). CAS: 115104-28-4
SDP
Supplier:  Enzo Life Sciences BMLRA1090005
Encompass_Preferred
A potent, selective, orally active leukotriene D4 antagonist. Ki=2.1 nM for inhibition of [3H]-LTD4 binding to human lung membranes. Competitively antagonizes contractions of guinea pig trachea and ileum and human trachea induced by LTD4. Blocks LTD4 and ascaris-induced bronchoconstriction in conscious squirrel monkeys. Alternative name: L660 711. Purity: ≥90% (HPLC). Solubility: Soluble in water (20mg/ml), aqueous buffers, 100% ethanol (10mg/ml) or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1712