Learn More
Medchemexpress LLC HY-10680 100mg , MK-6892 CAS:917910-45-3 Purity:98%
SDP
Catalog No. 501493285
Encompass_Preferred
Medchemexpress, HY-10680 100mg MK-6892 CAS:917910-45-3 Formula:C19H22N4O5 Ki: 4 nM (GPR109A) EC50: 16 nM (GPR109A) Purity:98% MK-6892 is a potent, selective, and full agonist for the high affinity nicotinic acid (NA) receptor GPR109A . K i and GTPγS EC 50 of MK-6892 on the Human GPR109A is 4 nM and 16 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3285 Supplier Medchemexpress LLC Supplier No. HY10680100MG
Medchemexpress, HY-10680 100mg MK-6892 CAS:917910-45-3 Formula:C19H22N4O5 Ki: 4 nM (GPR109A) EC50: 16 nM (GPR109A) Purity:98% MK-6892 is a potent, selective, and full agonist for the high affinity nicotinic acid (NA) receptor GPR109A . K i and GTPγS EC 50 of MK-6892 on the Human GPR109A is 4 nM and 16 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.