Learn More
Medchemexpress LLC Mk-761 free base | 65321-41-7 | 99.6% | 249.31 g/mol | C13H19N3O2 | 1MG
SDP
Catalog No. 5000238454
Encompass_Preferred
MK-761 free base is a research-grade β2-adrenergic blocker described as a potent, orally active compound with antihypertensive, positive inotropic, and vasodilating effects. It is supplied as the free base for laboratory use and is suitable for preclinical pharmacology and cardiovascular research.
  • Potent β2-adrenergic blocker suitable for receptor pharmacology studies.
  • Orally active profile observed in preclinical models.
  • Exhibits antihypertensive and positive inotropic effects.
  • Shows vasodilating properties in vitro and in vivo models.
  • High purity, typically around 99.6%.
  • Molecular formula C13H19N3O2 and molecular weight 249.31 g/mol.
Catalog No. 50-002-38454 Supplier Medchemexpress LLC Supplier No. HY1234491MG
MK-761 free base is a research-grade β2-adrenergic blocker described as a potent, orally active compound with antihypertensive, positive inotropic, and vasodilating effects. It is supplied as the free base for laboratory use and is suitable for preclinical pharmacology and cardiovascular research.
  • Potent β2-adrenergic blocker suitable for receptor pharmacology studies.
  • Orally active profile observed in preclinical models.
  • Exhibits antihypertensive and positive inotropic effects.
  • Shows vasodilating properties in vitro and in vivo models.
  • High purity, typically around 99.6%.
  • Molecular formula C13H19N3O2 and molecular weight 249.31 g/mol.