Learn More
Medchemexpress LLC Mk-761 tfa | 99.9% | 363.33 | 25 MG
SDP
Catalog No. 5000438427
Encompass_Preferred
MK-761 TFA is a potent and orally active β2-adrenergic blocker. It shows antihypertensive and positive inotropic effects, as well as vasodilating properties. This product is for research use only.
  • Potent and orally active β2-adrenergic blocker
  • Shows antihypertensive effects
  • Shows positive inotropic effects
  • Exhibits vasodilating properties
Catalog No. 50-004-38427 Supplier Medchemexpress LLC Supplier No. 25MGHY123449A
MK-761 TFA is a potent and orally active β2-adrenergic blocker. It shows antihypertensive and positive inotropic effects, as well as vasodilating properties. This product is for research use only.
  • Potent and orally active β2-adrenergic blocker
  • Shows antihypertensive effects
  • Shows positive inotropic effects
  • Exhibits vasodilating properties