Learn More
Medchemexpress LLC MK-761 TFA | 99.9% | 363.33 g/mol | C15H20F3N3O4 | 5 MG
SDP
Catalog No. 5000264447
Encompass_Preferred
MK-761 TFA is a potent, orally active β2-adrenergic blocker with antihypertensive, positive inotropic, and vasodilating effects. It is supplied as the trifluoroacetate (TFA) salt as a solid, white to off-white material for research use.
  • Potent, orally active β2-adrenergic blocker.
  • Shows antihypertensive activity.
  • Exhibits positive inotropic effects.
  • Demonstrates vasodilating properties.
  • Supplied as the trifluoroacetate (TFA) salt.
  • High purity: 99.93%.
  • Available in small research pack sizes including 5 mg.
  • Storage: -20°C sealed for long-term, in solvent -80°C for up to 6 months.
Catalog No. 50-002-64447 Supplier Medchemexpress LLC Supplier No. HY123449A5MG
MK-761 TFA is a potent, orally active β2-adrenergic blocker with antihypertensive, positive inotropic, and vasodilating effects. It is supplied as the trifluoroacetate (TFA) salt as a solid, white to off-white material for research use.
  • Potent, orally active β2-adrenergic blocker.
  • Shows antihypertensive activity.
  • Exhibits positive inotropic effects.
  • Demonstrates vasodilating properties.
  • Supplied as the trifluoroacetate (TFA) salt.
  • High purity: 99.93%.
  • Available in small research pack sizes including 5 mg.
  • Storage: -20°C sealed for long-term, in solvent -80°C for up to 6 months.