Learn More
Abcam MK-7655, 5MG
Supplier:  Abcam AB2869885MG
Encompass_Preferred
MW 348.38 Da, Purity >=98%. Potent and selective beta-lactamase inhibitor. It restores imipenem-susceptibility in imipenem-resistant K. pneumoniae and P. aeruginosa when used at concentrations of 12.5 and 4.7 μM, respectively.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5855