Learn More
Apexbio Technology LLC (+)-MK 801(Synonyms: Dizocilpine, Dizocilpine maleate, MK-801, (+)-MK-801, (+)-Dizocilpine), 10mg, CAS: 70449-94-4.
SDP
Catalog No. 5000059997
Encompass_Preferred
( )-MK 801 (CAS 70449-94-4) is a potent selective and non-competitive antagonist of the N-methyl-D-aspartate (NMDA) receptor displaying a Ki of approximately 30 5 nM This small molecule readily crosses the blood-brain barrier and interacts reversibly with specific binding sites on cortical membranes predominantly localized within the hippocampus In vitro assays utilizing rat cortical slice preparations indicate that ( )-MK 801 robustly inhibits NMDA-induced depolarization events and suppresses epileptiform activity mediated by neurotoxic agents Due to these pharmacological properties ( )-MK 801 is utilized experimentally in neuroscience research to study NMDA receptor-mediated events neurotoxicity convulsions and excitatory neurotransmission pathways
Catalog No. 50-000-59997 Supplier Apexbio Technology LLC Supplier No. A310010
( )-MK 801 (CAS 70449-94-4) is a potent selective and non-competitive antagonist of the N-methyl-D-aspartate (NMDA) receptor displaying a Ki of approximately 30 5 nM This small molecule readily crosses the blood-brain barrier and interacts reversibly with specific binding sites on cortical membranes predominantly localized within the hippocampus In vitro assays utilizing rat cortical slice preparations indicate that ( )-MK 801 robustly inhibits NMDA-induced depolarization events and suppresses epileptiform activity mediated by neurotoxic agents Due to these pharmacological properties ( )-MK 801 is utilized experimentally in neuroscience research to study NMDA receptor-mediated events neurotoxicity convulsions and excitatory neurotransmission pathways