Learn More
Apexbio Technology LLC (-)-MK 801 121917-57-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062007
Encompass_Preferred
(-)-MK 801 (CAS 121917-57-5) is a small molecule serving as a non-competitive antagonist at the N-methyl-D-aspartate (NMDA) receptor It crosses the blood-brain barrier binding reversibly and with high affinity to cortical membranes in a saturable regionally specific manner notably within the hippocampus (-)-MK 801 selectively inhibits depolarization responses to NMDA receptor activation leading to suppression of seizure-like neuronal activity induced by neurotoxins such as tetrodotoxin Due to these properties it has widespread use as a research tool in neuroscience particularly in investigating excitotoxicity epilepsy and neurological disorders associated with NMDA receptor dysfunction
Catalog No. 50-000-62007 Supplier Apexbio Technology LLC Supplier No. B16275.1
(-)-MK 801 (CAS 121917-57-5) is a small molecule serving as a non-competitive antagonist at the N-methyl-D-aspartate (NMDA) receptor It crosses the blood-brain barrier binding reversibly and with high affinity to cortical membranes in a saturable regionally specific manner notably within the hippocampus (-)-MK 801 selectively inhibits depolarization responses to NMDA receptor activation leading to suppression of seizure-like neuronal activity induced by neurotoxins such as tetrodotoxin Due to these properties it has widespread use as a research tool in neuroscience particularly in investigating excitotoxicity epilepsy and neurological disorders associated with NMDA receptor dysfunction