Learn More
Selleck Chemical LLC (-)-MK 801 maleate 10mg 121917-57-5 C13737
SDP
Supplier:  Selleck Chemical LLC S285710MG
Encompass_Preferred
(-)-MK-801 (Dizocilpine, C13737) is a potent N-methyl-D-aspartate (NMDA) receptor antagonist with Ki of 30.5 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3071