Learn More
Apexbio Technology LLC (+)-MK 801 Maleate, 50mg. Cas: 77086-22-7 MFCD: MFCD00082465. Potent NMDA antagonist
SDP
Supplier:  Apexbio Technology LLC A889650
Encompass_Preferred
(+)-MK 801 is a potent antagonist of NMDA with Ki value of 30.5nM [1].MK 801 is a potent anticonvulsant exhibits both anxiolytic and sympathomimetic properties. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5196