Learn More
Cayman Chemical MK-801maleate 10mg
SDP
Catalog No. 502940502
Encompass_Preferred
An NMDA receptor antagonist (Ki = 43 nM for inhibition of spermine binding); blocks generalized seizures in a mouse model of epilepsy (ED50 = 0.85 mg/kg); increases horizontal locomotor activity in mice (ED50 = 2.71 mg/kg); decreases immobility time in the tail suspension and forced swim tests in a mouse model of social defeat stress at 0.1 mg/kg; inhibits AChE (Kis = 6.2 and 7.9 μM for electric eel and rat brain AChE, respectively)
Catalog No. 50-294-0502 Supplier Cayman Chemical Supplier No. 2721310 MG
An NMDA receptor antagonist (Ki = 43 nM for inhibition of spermine binding); blocks generalized seizures in a mouse model of epilepsy (ED50 = 0.85 mg/kg); increases horizontal locomotor activity in mice (ED50 = 2.71 mg/kg); decreases immobility time in the tail suspension and forced swim tests in a mouse model of social defeat stress at 0.1 mg/kg; inhibits AChE (Kis = 6.2 and 7.9 μM for electric eel and rat brain AChE, respectively)