Learn More
Medchemexpress LLC MK-8033 hydrochloride | 1283000-43-0 | MFCD18633221 | ≥98.0% | C24H22Cl2N6O | 10MG
SDP
Catalog No. 5000260722
Encompass_Preferred
MK-8033 hydrochloride is a small-molecule, ATP-competitive dual inhibitor of c-Met and Ron kinases supplied as the hydrochloride salt for research use. It has been used in oncology research and was evaluated in early clinical trials; supplier listings report high purity and analytical characterization for laboratory applications.
  • Potent dual c-Met and Ron kinase inhibitor
  • Hydrochloride salt form
  • Purity ≥98.0%
  • Characterized by molecular formula and molecular weight
  • Intended for research use in oncology studies
  • Documented in early-phase clinical investigations
Catalog No. 50-002-60722 Supplier Medchemexpress LLC Supplier No. HY13299A10MG
MK-8033 hydrochloride is a small-molecule, ATP-competitive dual inhibitor of c-Met and Ron kinases supplied as the hydrochloride salt for research use. It has been used in oncology research and was evaluated in early clinical trials; supplier listings report high purity and analytical characterization for laboratory applications.
  • Potent dual c-Met and Ron kinase inhibitor
  • Hydrochloride salt form
  • Purity ≥98.0%
  • Characterized by molecular formula and molecular weight
  • Intended for research use in oncology studies
  • Documented in early-phase clinical investigations