Learn More
TARGETMOL CHEMICALS INC MK-8245 10MG
SDP
Catalog No. 502580694
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-8245 is a liver-targeting SCD inhibitor for human SCD1 (IC50 1 nM) and for rat/mouse SCD1 (IC50 3 nM) with anti-diabetic and anti-dyslipidemic function. purity: 99%
Catalog No. 50-258-0694 Supplier TARGETMOL CHEMICALS INC Supplier No. T265010MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-8245 is a liver-targeting SCD inhibitor for human SCD1 (IC50 1 nM) and for rat/mouse SCD1 (IC50 3 nM) with anti-diabetic and anti-dyslipidemic function. purity: 99%