Learn More
Apexbio Technology LLC MK-8245 1030612-90-8 200mg
SDP
Catalog No. 5000062254
Encompass_Preferred
MK-8245 (CAS 1030612-90-8) is a potent liver-selective small molecule inhibitor of stearoyl-CoA desaturase (SCD) SCD1 catalyzes fatty acid desaturation and represents a target for treating type II diabetes obesity dyslipidemia and other metabolic disorders MK-8245 demonstrates strong inhibitory activity against rat mouse and human SCD1 with an IC50 of approximately 1 nM and displays liver-specific tissue distribution mediated by organic anion transporting polypeptides (OATPs) Animal studies revealed MK-8245 reduces blood glucose levels and hepatic triglycerides dose-dependently supporting its application in metabolic disease research
Catalog No. 50-000-62254 Supplier Apexbio Technology LLC Supplier No. A4345200
MK-8245 (CAS 1030612-90-8) is a potent liver-selective small molecule inhibitor of stearoyl-CoA desaturase (SCD) SCD1 catalyzes fatty acid desaturation and represents a target for treating type II diabetes obesity dyslipidemia and other metabolic disorders MK-8245 demonstrates strong inhibitory activity against rat mouse and human SCD1 with an IC50 of approximately 1 nM and displays liver-specific tissue distribution mediated by organic anion transporting polypeptides (OATPs) Animal studies revealed MK-8245 reduces blood glucose levels and hepatic triglycerides dose-dependently supporting its application in metabolic disease research