Learn More
Selleck Chemical LLC MK-8245 5mg 1030612-90-8
SDP
Supplier:  Selleck Chemical LLC S11585MG
Encompass_Preferred
MK-8245 is an liver-targeting inhibitor of stearoyl-CoA desaturase (SCD) with IC50 of 1 nM for human SCD1 and 3 nM for both rat SCD1 and mouse SCD1, with anti-diabetic and anti-dyslipidemic efficacy. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0965