Learn More
Apexbio Technology LLC MK-8245, 5mg. Cas: 1030612-90-8 MFCD: MFCD20039622. SCD inhibitor,potent and liver-selective
SDP
Supplier:  Apexbio Technology LLC A43455
Encompass_Preferred
MK-8245 is a potent and liver-selective inhibitor of stearoyl-CoA desaturase (SCD) with IC50 value of 1nM [1].SCD1 represents a therapeutic target for the treatment of type II diabetes, dyslipidemia, obesity, and metabolic diseases. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4927