Learn More
Medchemexpress LLC MK-8245 Trifluoroacetate | 1415559-41-9 | 98.5% | 1 MG
SDP
Catalog No. 5000339820
Encompass_Preferred
MK-8245 trifluoroacetate is a liver-targeting inhibitor of stearoyl-CoA desaturase (SCD) with anti-diabetic and anti-dyslipidemic efficacy. It demonstrates high potency with an IC50 of 1 nM for human SCD1 and 3 nM for rat and mouse SCD1, and selectively targets Δ-5 and Δ-6 desaturases. Its tetrazole acetic acid moiety facilitates OATPs recognition and liver-targeting.
  • Liver-targeting inhibitor of stearoyl-CoA desaturase (SCD)
  • Potent inhibition with IC50 of 1 nM for human SCD1
  • Shows anti-diabetic and anti-dyslipidemic efficacy
  • Tetrazole acetic acid moiety aids OATPs recognition and liver-targeting
  • Exhibits significant SCD inhibition in rat hepatocyte assay (IC50 of 68 nM)
  • Displays highly selective activity for Δ-5 and Δ-6 desaturases
  • Concentrated in the liver in vivo, with low exposure in other tissues
  • Improves glucose clearance in eDIO mice
Catalog No. 50-003-39820 Supplier Medchemexpress LLC Supplier No. HY130771MG
MK-8245 trifluoroacetate is a liver-targeting inhibitor of stearoyl-CoA desaturase (SCD) with anti-diabetic and anti-dyslipidemic efficacy. It demonstrates high potency with an IC50 of 1 nM for human SCD1 and 3 nM for rat and mouse SCD1, and selectively targets Δ-5 and Δ-6 desaturases. Its tetrazole acetic acid moiety facilitates OATPs recognition and liver-targeting.
  • Liver-targeting inhibitor of stearoyl-CoA desaturase (SCD)
  • Potent inhibition with IC50 of 1 nM for human SCD1
  • Shows anti-diabetic and anti-dyslipidemic efficacy
  • Tetrazole acetic acid moiety aids OATPs recognition and liver-targeting
  • Exhibits significant SCD inhibition in rat hepatocyte assay (IC50 of 68 nM)
  • Displays highly selective activity for Δ-5 and Δ-6 desaturases
  • Concentrated in the liver in vivo, with low exposure in other tissues
  • Improves glucose clearance in eDIO mice