Learn More
TARGETMOL CHEMICALS INC MK-8325 5MG
SDP
Catalog No. 502580802
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-8325 is a potent and orally available HCV NS5A inhibitor with replicative activity against a wide range of genotypes.MK-8325 has demonstrated bioavailability in in vitro and in vivo assays and has a favorable overall ADME profile. purity: 98%
Catalog No. 50-258-0802 Supplier TARGETMOL CHEMICALS INC Supplier No. T244805MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-8325 is a potent and orally available HCV NS5A inhibitor with replicative activity against a wide range of genotypes.MK-8325 has demonstrated bioavailability in in vitro and in vivo assays and has a favorable overall ADME profile. purity: 98%