Learn More
Medchemexpress LLC MK-8353 | 1184173-73-6 | MFCD31657387 | 98.1% | 691.84 | 50 MG
SDP
Catalog No. 5000344121
Encompass_Preferred
MK-8353 (SCH900353) is a potent, selective, and orally available ERK1/2 inhibitor with demonstrated antitumor activity. It functions as an EC 2.7.11.24 (mitogen-activated protein kinase) inhibitor, an antineoplastic agent, and an apoptosis inducer.
  • Potent and selective ERK1/2 inhibitor
  • Orally available
  • Exhibits antitumor activity
  • IC50s of 23.0 nM for ERK1 and 8.8 nM for ERK2
  • Functions as a mitogen-activated protein kinase inhibitor
  • Acts as an antineoplastic agent
  • Induces apoptosis
Catalog No. 50-003-44121 Supplier Medchemexpress LLC Supplier No. HY11140750MG
MK-8353 (SCH900353) is a potent, selective, and orally available ERK1/2 inhibitor with demonstrated antitumor activity. It functions as an EC 2.7.11.24 (mitogen-activated protein kinase) inhibitor, an antineoplastic agent, and an apoptosis inducer.
  • Potent and selective ERK1/2 inhibitor
  • Orally available
  • Exhibits antitumor activity
  • IC50s of 23.0 nM for ERK1 and 8.8 nM for ERK2
  • Functions as a mitogen-activated protein kinase inhibitor
  • Acts as an antineoplastic agent
  • Induces apoptosis