Learn More
Medchemexpress LLC HY-111407 5mg Medchemexpress, MK-8353 CAS:1184173-73-6 Purity:>98%
SDP
Catalog No. 501968859
Encompass_Preferred
Medchemexpress, HY-111407 5mg MK-8353 CAS:1184173-73-6 MK-8353 (SCH900353) is a potent, selective and orally available ERK1/2 inhibitor, with IC50s of 23.0 nM and 8.8 nM, respectively; MK-8353 has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8859 Supplier Medchemexpress LLC Supplier No. HY1114075MG
Medchemexpress, HY-111407 5mg MK-8353 CAS:1184173-73-6 MK-8353 (SCH900353) is a potent, selective and orally available ERK1/2 inhibitor, with IC50s of 23.0 nM and 8.8 nM, respectively; MK-8353 has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.