Learn More
Selleck Chemical LLC MK-8353 (SCH900353) 2mg 1184173-73-6
SDP
Supplier:  Selleck Chemical LLC S87012MG
Encompass_Preferred
MK-8353 (SCH900353) is an orally bioavailable, selective, and potent ERK inhibitor that inhibits activated ERK1 and ERK2 in vitro, with IC50 values of 23.0 nM and 8.8 nM, respectively (IMAP kinase assay), and nonactivated ERK2, with an IC50 of 0.5 nM (MEK1-ERK2-coupled assay). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8213