Learn More
Selleck Chemical LLC MK-8617 25mg 1187990-87-9
SDP
Supplier:  Selleck Chemical LLC S844325MG
Encompass_Preferred
MK-8617 is an orally active pan-inhibitor of Hypoxia-inducible factor prolyl hydroxylase 1?3 (HIF PHD1?3), inhibiting PHD1, 2, 3 with IC50s of 1.0, 1.0 and 14 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6065