Learn More
Apexbio Technology LLC MK-8745 885325-71-3 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A88075.1
Encompass_Preferred
MK-8745 (CAS 885325-71-3) is a small-molecule inhibitor targeting Aurora A kinase a serine/threonine kinase regulating chromosomal segregation during mitosis Aberrant Aurora A activity correlates with cancer progression In preclinical studies MK-8745 suppresses Aurora A kinase eliciting p53-dependent apoptosis and G2/M cell cycle arrest MK-8745 demonstrates antitumor activity in vitro across various cancer cell lines including p53-deficient and checkpoint-defective HCT116 variants and inhibits tumor growth in xenografted tumor models signifying its potential utility in cancer research targeting Aurora kinase pathways
Catalog No. 50-000-63361