Learn More
Apexbio Technology LLC MK-8776 (SCH-900776) 891494-63-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A84775.1
Encompass_Preferred
MK-8776 (SCH-900776 CAS number 891494-63-6) is a selective small molecule inhibitor targeting the serine/threonine kinase checkpoint kinase 1 (Chk1) Chk1 is involved in detecting DNA damage and regulating DNA replication particularly in stabilizing replication forks during genotoxic stress SCH-900776 selectively inhibits Chk1 activity (IC50 3 nM) over Chk2 and Cdk3 In cellular assays SCH-900776 treatment results in dose-dependent inhibition of Chk1 autophosphorylation at Ser296 accompanied by increased phosphorylation at Ser345 a cellular marker consistent with checkpoint disruption In preclinical models combining SCH-900776 with DNA-damaging agents such as gemcitabine enhances anti-tumor efficacy marking its potential utility in cancer research
Catalog No. 50-000-61845