Learn More
Selleck Chemical LLC MK-8776 (SCH 900776) 5mg 891494-63-6
SDP
Supplier:  Selleck Chemical LLC S27355MG
Encompass_Preferred
MK-8776 (SCH 900776) is a selective Chk1 inhibitor with IC50 of 3 nM in a cell-free assay. It shows 500-fold selectivity against Chk2. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0951094