Learn More
Medchemexpress LLC HY-14166 5mg Medchemexpress, MK-886 CAS:118414-82-7 Purity:>98%
SDP
Catalog No. 502030515
Encompass_Preferred
Medchemexpress, HY-14166 5mg MK-886 CAS:118414-82-7 MK-886 (L 663536) is a potent, cell-permeable and orally active FLAP (IC50 of 30 nM) and leukotriene biosynthesis (IC50s of 3 nM and 1.1 μM in intact leukocytes and human whole blood, respectively) inhibitor. MK-886 is also a non-competitive PPARα antagonist and can induce apoptosis[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0515 Supplier Medchemexpress LLC Supplier No. HY141665MG
Medchemexpress, HY-14166 5mg MK-886 CAS:118414-82-7 MK-886 (L 663536) is a potent, cell-permeable and orally active FLAP (IC50 of 30 nM) and leukotriene biosynthesis (IC50s of 3 nM and 1.1 μM in intact leukocytes and human whole blood, respectively) inhibitor. MK-886 is also a non-competitive PPARα antagonist and can induce apoptosis[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.