Learn More
Selleck Chemical LLC MK-886
SDP
Supplier:  Selleck Chemical LLC S82365MG
Encompass_Preferred
MK-886 is an inhibitor of leukotriene biosynthesis inhibiting 5-lipoxygenase-activating protein (FLAP) It is also a moderately potent PPAR antagonist
Catalog No. 50-136-5828