Learn More
American Radiolabeled Chemicals Inc MK 912 3H 250UCMK 912 3H 2
SDP
Catalog No. 50901103
Encompass
MK-912-methyl-3H 250uCi in ethanol, 1 mCi/ml, S.A. 60-87 Ci/mmol, Shipped in Dry Ice
Catalog No. 50-901-103 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1753250 UC
Add to Cart
Add to Cart
Request a Quote
MK-912-methyl-3H 250uCi in ethanol, 1 mCi/ml, S.A. 60-87 Ci/mmol, Shipped in Dry Ice