Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MK2-IN-1 hydrochloride | 1314118-94-9 | 98.2% | 50 MG
SDP

Catalog No. 5000342501
Encompass_Preferred

MK2-IN-1 hydrochloride (compound 1) is a potent and selective MAPKAPK2 (MK2) inhibitor. It features an IC50 of 0.11 uM for MK2 and an EC50 of 0.35 uM for pHSP27, and impairs the phosphorylation level of serine residues in the Tfcp2l1 protein.

  • Increases Tfcp2l1 protein level without changing transcript level within 2 hours
  • Induces alkaline phosphatase (AP)-positive colonies in short time
  • Soluble in H2O at ≥ 100 mg/mL
  • Soluble in DMSO at 100 mg/mL (requires ultrasonication)
  • Appearance: Solid
  • Color: Light yellow to pink

Catalog No. 50-003-42501 Supplier Medchemexpress LLC Supplier No. HY12834A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MK2-IN-1 hydrochloride (compound 1) is a potent and selective MAPKAPK2 (MK2) inhibitor. It features an IC50 of 0.11 uM for MK2 and an EC50 of 0.35 uM for pHSP27, and impairs the phosphorylation level of serine residues in the Tfcp2l1 protein.

  • Increases Tfcp2l1 protein level without changing transcript level within 2 hours
  • Induces alkaline phosphatase (AP)-positive colonies in short time
  • Soluble in H2O at ≥ 100 mg/mL
  • Soluble in DMSO at 100 mg/mL (requires ultrasonication)
  • Appearance: Solid
  • Color: Light yellow to pink

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.