Learn More
Medchemexpress LLC MK2-IN-1 (hydrochloride) | 1314118-94-9 | 98.2% | 509.43 | 10 MG
SDP
Catalog No. 5000365065
Encompass_Preferred
MK2-IN-1 hydrochloride is a potent and selective MAPKAPK2 (MK2) inhibitor with an IC50 of 0.11 μM for MK2 and an EC50 of 0.35 μM for pHSP27. It impairs the phosphorylation level of serine residues in the Tfcp2l1 protein, making it suitable for studies on protein regulation and cell signaling pathways.
  • Increases Tfcp2l1 protein level in mouse embryonic stem cells.
  • Induces alkaline phosphatase positive colonies.
  • Suitable for in vitro studies with various dissolution protocols.
Catalog No. 50-003-65065 Supplier Medchemexpress LLC Supplier No. 10MGHY12834A
MK2-IN-1 hydrochloride is a potent and selective MAPKAPK2 (MK2) inhibitor with an IC50 of 0.11 μM for MK2 and an EC50 of 0.35 μM for pHSP27. It impairs the phosphorylation level of serine residues in the Tfcp2l1 protein, making it suitable for studies on protein regulation and cell signaling pathways.
  • Increases Tfcp2l1 protein level in mouse embryonic stem cells.
  • Induces alkaline phosphatase positive colonies.
  • Suitable for in vitro studies with various dissolution protocols.