Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MK2-IN-1 (hydrochloride) | 1314118-94-9 | 98.4% | 1 ML
SDP

Catalog No. 5000339073
Encompass_Preferred

MK2-IN-1 hydrochloride (compound 1) is a potent and selective MAPKAPK2 (MK2) inhibitor with an IC50 of 0.11 μM for MK2 and an EC50 of 0.35 μM for pHSP27. It impairs the phosphorylation level of serine residues in the Tfcp2l1 protein. This product is for research use only and not sold to patients.

  • Potent and selective MAPKAPK2 (MK2) inhibitor
  • Gradually increases Tfcp2l1 protein level without a change in transcript level within 2 hours
  • Induces more alkaline phosphatase (AP)-positive colonies
  • Molecular weight of 509.43
  • Chemical formula C27H26Cl2N4O2
  • Appears as a light yellow to pink solid
  • Highly soluble in H2O and DMSO

Catalog No. 50-003-39073 Supplier Medchemexpress LLC Supplier No. HY12834A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MK2-IN-1 hydrochloride (compound 1) is a potent and selective MAPKAPK2 (MK2) inhibitor with an IC50 of 0.11 μM for MK2 and an EC50 of 0.35 μM for pHSP27. It impairs the phosphorylation level of serine residues in the Tfcp2l1 protein. This product is for research use only and not sold to patients.

  • Potent and selective MAPKAPK2 (MK2) inhibitor
  • Gradually increases Tfcp2l1 protein level without a change in transcript level within 2 hours
  • Induces more alkaline phosphatase (AP)-positive colonies
  • Molecular weight of 509.43
  • Chemical formula C27H26Cl2N4O2
  • Appears as a light yellow to pink solid
  • Highly soluble in H2O and DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.