Learn More
TARGETMOL CHEMICALS INC MK2-IN-1 HYDROCHLORIDE 25MG
SDP
Catalog No. 502579882
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK2-IN-1 hydrochloride (MK 25) is a highly selective non-ATP competitive inhibitor of p38/mitogen-activated protein kinase-activated protein kinase 2 (MAPKAPK2 or MK2 IC50 0.11 uM) [1]. purity: 99%
Catalog No. 50-257-9882 Supplier TARGETMOL CHEMICALS INC Supplier No. T444225MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK2-IN-1 hydrochloride (MK 25) is a highly selective non-ATP competitive inhibitor of p38/mitogen-activated protein kinase-activated protein kinase 2 (MAPKAPK2 or MK2 IC50 0.11 uM) [1]. purity: 99%