Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mk2-in-1 hydrochloride | 1314118-94-9 | 98.4% | 509.43 g/mol | C27H26Cl2N4O2 | 5 MG
SDP

Catalog No. 501872744
Encompass_Preferred

MK2-IN-1 hydrochloride is the hydrochloride salt of a small-molecule MAPKAPK2 (MK2) inhibitor intended for biochemical and cell-based research. It provides potent biochemical activity with high solubility and is supplied as a solid for laboratory use.

  • Potent and selective MAPKAPK2 (MK2) inhibitor (IC50 = 0.11 μM; EC50 = 0.35 μM).
  • High purity (98.36%).
  • Molecular formula C27H26Cl2N4O2; molecular weight 509.43 g/mol.
  • Solid, light yellow to pink appearance suitable for handling and storage.
  • High solubility in water and DMSO (water ≥ 100 mg/mL; DMSO 100 mg/mL).
  • Includes recommended in vivo formulation options for dosing studies.

Catalog No. 50-187-2744 Supplier Medchemexpress LLC Supplier No. HY12834A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MK2-IN-1 hydrochloride is the hydrochloride salt of a small-molecule MAPKAPK2 (MK2) inhibitor intended for biochemical and cell-based research. It provides potent biochemical activity with high solubility and is supplied as a solid for laboratory use.

  • Potent and selective MAPKAPK2 (MK2) inhibitor (IC50 = 0.11 μM; EC50 = 0.35 μM).
  • High purity (98.36%).
  • Molecular formula C27H26Cl2N4O2; molecular weight 509.43 g/mol.
  • Solid, light yellow to pink appearance suitable for handling and storage.
  • High solubility in water and DMSO (water ≥ 100 mg/mL; DMSO 100 mg/mL).
  • Includes recommended in vivo formulation options for dosing studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.