Learn More
Medchemexpress LLC MK2-IN-3 hydrate | 1186648-22-5 | 99.1% | 358.39 | 50 MG
SDP
Catalog No. 5000436880
Encompass_Preferred
MK2-IN-3 hydrate (compound 16) is an orally active, selective, and ATP-competitive MAPKAP-K2 (MK-2) inhibitor with an IC50 of 8.5 nM. It exhibits exceptional selectivity against MK-3, MK-5, ERK2, and MNK1, as well as CDK2, JNK2, IKK2, MSK1, and MSK2. For research use only.
  • Suppresses TNFα production in U397 cells with an IC50 of 4.4 μM
  • Orally active and selective
  • ATP-competitive inhibitor
Catalog No. 50-004-36880 Supplier Medchemexpress LLC Supplier No. HY11245750MG
MK2-IN-3 hydrate (compound 16) is an orally active, selective, and ATP-competitive MAPKAP-K2 (MK-2) inhibitor with an IC50 of 8.5 nM. It exhibits exceptional selectivity against MK-3, MK-5, ERK2, and MNK1, as well as CDK2, JNK2, IKK2, MSK1, and MSK2. For research use only.
  • Suppresses TNFα production in U397 cells with an IC50 of 4.4 μM
  • Orally active and selective
  • ATP-competitive inhibitor