Learn More
Selleck Chemical LLC MK2 Inhibitor III
SDP
Supplier:  Selleck Chemical LLC S69301G
Encompass_Preferred
MK2 Inhibitor III (MK2-IN-3) is an ATP-competitive MK2 (MAPKAP-K2) inhibitor with an IC50 value of 8 5 nM
Catalog No. 50-000-02240
Add to Cart
Request a Quote