Learn More
Cayman Chemical MK2 InhIbItr IV 5mg
SDP
Catalog No. 502970063
Encompass_Preferred
A highly selective, non-ATP competitive inhibitor of p38/mitogen-activated protein kinase-activated protein kinase 2 (IC50 = 0.11 μM); inhibits TNF-α (IC50 = 4.4 μM) and IL-6 (IC50 = 5.2 μM) secretion from the human THP-1 acute monocytic leukemia cell line
Catalog No. 50-297-0063 Supplier Cayman Chemical Supplier No. 143995 MG
A highly selective, non-ATP competitive inhibitor of p38/mitogen-activated protein kinase-activated protein kinase 2 (IC50 = 0.11 μM); inhibits TNF-α (IC50 = 4.4 μM) and IL-6 (IC50 = 5.2 μM) secretion from the human THP-1 acute monocytic leukemia cell line