Learn More
Proteintech Group Inc MK2-Trap Agarose, kit
SDP
Supplier:  Proteintech Group Inc MTAK20
Encompass
ChromoTek’s MK2-Trap Agarose kit are affinity beads for IP of MK2-fusion proteins. It comprises a MK2 Nanobody/ VHH conjugated to agarose beads.
Catalog No. 50-225-8536