Learn More
Mopec MK200 Elevating Down Draft Grossing Station
SDP
Catalog No. 22444214 Shop All Mopec Products
Change view
Click to view available options
Height (Metric):
81.3 cm to 101.6 cm
Catalog No. Height (Metric)
22-444-214 81.3 cm to 101.6 cm
1 options
Catalog No. 22-444-214 Supplier Mopec Supplier No. MK200
Designed with a grid plate work surface that elevates to the most agreeable elevation to perform duties without back strain. Mopec™ MK200Elevating Down Draft Grossing Station is an elevating, two sided, ventilated grossing workstation is designed to allow technician access to either side.
  • Wrist blade handle controls are not available with this option
  • Must be facility installed on the ME200. Shipped loose with ME200
  • M0039 is not compatible with this option
Warranty and Services
1 Year Warranty
Specifications
Product Type Grossing Station
Includes Dual foot pedal H/C water control valve and a swing spout faucet.
Height (Metric) 81.3 cm to 101.6 cm