Learn More
Cayman Chemical MK2a Inhibitor, Purity: ≥98%, CAS: 41179-33-3, 50 mg
SDP
Catalog No. 502965760
Encompass_Preferred
A substrate-selective p38α MAPK inhibitor; selectively inhibits p38α-dependent phosphorylation of MK2 over ATF2 (Ki = 330 and >20,000 nM, respectively, in cell-free assays); inhibits proliferation of U87, A172, and U251 glioblastoma cells (EC50s = 0.6-1 μM) via inhibition of tubulin polymerization and induction of apoptosis
Catalog No. 50-296-5760 Supplier Cayman Chemical Supplier No. 1732550 MG
A substrate-selective p38α MAPK inhibitor; selectively inhibits p38α-dependent phosphorylation of MK2 over ATF2 (Ki = 330 and >20,000 nM, respectively, in cell-free assays); inhibits proliferation of U87, A172, and U251 glioblastoma cells (EC50s = 0.6-1 μM) via inhibition of tubulin polymerization and induction of apoptosis