Learn More
Medchemexpress LLC HY-10037 5mg Medchemexpress, MK591 (free acid) CAS:136668-42-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100375MG
Encompass_Preferred
Medchemexpress, HY-10037 5mg MK591 (free acid) CAS:136668-42-3 MK591 (free acid) is a selective and specific 5-Lipoxygenase-activating protein (FLAP) inhibitor with an IC 50 value of 1.6 nM in a FLAP binding assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1792