Learn More
Cayman Chemical (+)-MK-801 (hydrogen maleate), Purity: ≥98%, CAS: 77086-22-7, 10 mg
SDP
Catalog No. 502888967
Encompass_Preferred
Potent, selective and non-competitive NMDA receptor antagonist (Ki = 30.5 nM); acts at the NMDA receptor-operated ion channel as an open channel blocker, preventing Ca2+ flux
Catalog No. 50-288-8967 Supplier Cayman Chemical Supplier No. 1000901910 MG
Potent, selective and non-competitive NMDA receptor antagonist (Ki = 30.5 nM); acts at the NMDA receptor-operated ion channel as an open channel blocker, preventing Ca2+ flux