Learn More
Medchemexpress LLC HY-111363 10mg Medchemexpress, MK8722 CAS:1394371-71-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11136310MG
Encompass_Preferred
Medchemexpress, HY-111363 10mg MK8722 CAS:1394371-71-1 MK8722 is a potent and systemic pan-AMPK activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8842