Learn More
Abcam MK886, Lipo x ygenase inhibitor, 50MG
Catalog No. 502706855
Encompass_Preferred
MW 494.1 Da, Purity >98%. Potent 5-lipoxygenase activating protein inhibitor (FLAP) (IC₅₀ = 44 nM). Inhibits leukotriene biosynthesis (IC₅₀ = 2.5 nM) in vitro and tumor activity in vivo. Displays PPARα antagonist activity. Orally active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6855 Supplier Abcam Supplier No. AB14114050MG
MW 494.1 Da, Purity >98%. Potent 5-lipoxygenase activating protein inhibitor (FLAP) (IC₅₀ = 44 nM). Inhibits leukotriene biosynthesis (IC₅₀ = 2.5 nM) in vitro and tumor activity in vivo. Displays PPARα antagonist activity. Orally active in vivo.
The product is subject to the following: Abcam Restricted Use Statement