Learn More
Cayman Chemical MK-886 (sodium salt), Purity: ≥99%, CAS: 118427-55-7, 5 mg
SDP
Catalog No. 502891082
Encompass_Preferred
A potent FLAP antagonist that prevents 5-LO activation in vivo; inhibits LT biosynthesis in leukocytes with an IC50 value of 2.5 nM
Catalog No. 50-289-1082 Supplier Cayman Chemical Supplier No. 101335 MG
A potent FLAP antagonist that prevents 5-LO activation in vivo; inhibits LT biosynthesis in leukocytes with an IC50 value of 2.5 nM