Learn More
TARGETMOL CHEMICALS INC MKC-1 5MG
SDP
Catalog No. 502578952
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MKC-1 (Ro-31-7453) is an orally bioavailable small-molecule bisindolylmaleimide cell cycle inhibitor with potential antineoplastic activity. MKC-1 and its metabolites inhibit tubulin polymerization blocking the formation of the mitotic spindle which may result inpurity: 99%
Catalog No. 50-257-8952 Supplier TARGETMOL CHEMICALS INC Supplier No. T98315MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MKC-1 (Ro-31-7453) is an orally bioavailable small-molecule bisindolylmaleimide cell cycle inhibitor with potential antineoplastic activity. MKC-1 and its metabolites inhibit tubulin polymerization blocking the formation of the mitotic spindle which may result inpurity: 99%