Learn More
Selleck Chemical LLC MKC-3946 5mg 1093119-54-0
SDP
Supplier:  Selleck Chemical LLC S82865MG
Encompass_Preferred
MKC3946 is a potent and soluble IRE1? endoribonuclease domain inhibitor which triggered modest growth inhibition in multiple myeloma cell lines, without toxicity in normal mononuclear cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1523776