Learn More
Medchemexpress LLC HY-12399 1mg Medchemexpress, MKC9989 CAS:1338934-20-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY123991MG
Encompass_Preferred
Medchemexpress, HY-12399 1mg MKC9989 CAS:1338934-20-5 MKC9989 is a Hydroxy aryl aldehydes (HAA) inhibitor and also inhibits IRE1α with an IC50 of 0.23 to 44 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1034