Learn More
Medchemexpress LLC HY-137552 5mg Medchemexpress, MKI-1 CAS:1190277-80-5 Purity:>98%
SDP
Catalog No. 502081500
Encompass_Preferred
Medchemexpress, HY-137552 5mg MKI-1 CAS:1190277-80-5 MKI-1, an inhibitor of MASTL (microtubule-associated serine/threonine kinase-like) with an IC50 of 9.9 μM, exerts antitumor and radiosensitizer activities through PP2A activation in breast cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1500 Supplier Medchemexpress LLC Supplier No. HY1375525MG
Medchemexpress, HY-137552 5mg MKI-1 CAS:1190277-80-5 MKI-1, an inhibitor of MASTL (microtubule-associated serine/threonine kinase-like) with an IC50 of 9.9 μM, exerts antitumor and radiosensitizer activities through PP2A activation in breast cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.