Learn More
Medchemexpress LLC MKI-1 (MASTL Kinase Inhibitor-1) | 1190277-80-5 | 98.1% | 302.33 | 100 MG
SDP
Catalog No. 5000350692
Encompass_Preferred
MKI-1 is a novel small-molecule inhibitor of MASTL (microtubule-associated serine/threonine kinase-like) with an IC50 of 9.9 μM. It demonstrates antitumor and radiosensitizer activities by activating PP2A in breast cancer.
  • Inhibits MASTL activity.
  • Suppresses various oncogenic properties in cancer cells.
  • Reduces both serine 62-phosphorylation and total c-Myc, alongside a decrease in ENSA phosphorylation.
  • Decreases c-Myc stability through PP2A activation.
  • Reduces tumor growth and enhances radiosensitivity in xenograft models.
  • Exhibits low gross toxicity in treated animal models, with no notable changes in body weight.
Catalog No. 50-003-50692 Supplier Medchemexpress LLC Supplier No. HY137552100MG
MKI-1 is a novel small-molecule inhibitor of MASTL (microtubule-associated serine/threonine kinase-like) with an IC50 of 9.9 μM. It demonstrates antitumor and radiosensitizer activities by activating PP2A in breast cancer.
  • Inhibits MASTL activity.
  • Suppresses various oncogenic properties in cancer cells.
  • Reduces both serine 62-phosphorylation and total c-Myc, alongside a decrease in ENSA phosphorylation.
  • Decreases c-Myc stability through PP2A activation.
  • Reduces tumor growth and enhances radiosensitivity in xenograft models.
  • Exhibits low gross toxicity in treated animal models, with no notable changes in body weight.