Learn More
Eurofins Discoverx MKK6/SKK3 (S599D, T603D), active, 10 µg. (14-537)
Supplier:  Eurofins Discoverx 14537
Encompass
N-terminal MBP-tagged recombinant, human MKK6/SKK3 amino acids 4-end containing S599D and T603D mutations. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8939